
Ammonium hexachlororuthenate(IV) 99.99% (trace metals basis)
Catalog # TS43693-0010
Supplier: Thermo Scientific Chemicals
CAS Number:
Specifications
- Pack type:Glass bottle
- Size:1 g
- Cat. No.:TS43693-0010
Specifications
Specification Test Results
- Appearance:Dark brown to black Powder
- Total trace metal impurities:≤100 ppm
Specification Test Results
About this item
:
:349.86
Storage Temperature:Ambient
UN:3288
ADR:6.1,III
:GHS06
CAS Number:18746-63-9
Formula:H₈Cl₆N₂Ru
MDL Number:MFCD00015962