Order Entry
United States
ContactUsLinkComponent
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Irritant
Oxidizing
Environment
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Catalog # TS43710-0050
CAS Number:  
undefined
Irritant
Oxidizing
Environment
Chromium(III) nitrate nonahydrate 99.99% (trace metals basis)
Catalog # TS43710-0050
Supplier Number:  AC43710-0050
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    5 g
  • Cat. No.:
    TS43710-0050

Specifications

Specification Test Results

  • Appearance:
    Violet to black Crystals
  • Titration Iodometric:
    12.7 to 13.2 % (Cr)
  • Total trace metal impurities:
    ≤100 ppm

Specification Test Results

About this item

 :
 :400.15 g/mol
Melting Pt:60 °C
Storage Temperature:Ambient
UN:2720
ADR:5.1,III
 :GHS07
 :GHS03
 :GHS09
CAS Number:7789-02-8
Formula:CrN₃O₉·9H₂O
MDL Number:MFCD00149659