
Iron(II) sulfate heptahydrate 99+% ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:7782-63-0
:
Formula:FeSO₄·7H₂O
MDL Number:MFCD00149719
:278.02 g/mol
Merck Index:13,04091
Melting Pt:64 °C
EINECS:231-753-5
:GHS07
Storage Temperature:Ambient
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specification Test Results
- Appearance:Green to blue-green to light blue Crystals
- Titration with KMnO4:≥99.0 %
- Chloride (Cl):≤10 ppm
- Phosphate (PO4):≤10 ppm
- Calcium (Ca):≤50 ppm
- Copper (Cu):≤50 ppm
- Magnesium (Mg):≤20 ppm
- Manganese (Mn):≤500 ppm
- Potassium (K):≤20 ppm
- Sodium (Na):≤200 ppm
- Zinc (Zn):≤50 ppm
- Ferric iron (Fe 3+):≤1000 ppm (May increase due to inherent oxidation on storage)
- Insoluble matter:≤0.01 %
Specification Test Results
Frequently Bought Together