
1-Butanesulfonic acid sodium salt, anhydrous 99+% for ion pair chromatography
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:2386-54-1
:
Formula:C₄H₉NaO₃S
MDL Number:MFCD00007540
:160.17 g/mol
Melting Pt:>300 °C
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Infrared spectrum:Conforms
- Loss on drying:≤1 % (150°C)
- UV:(0.5 M in water)
- Titration after Ion exchange:≥99.0 % (on dry substance)
- Membrane filtration test:(on 0.5 M soln.) Passes test
- Suitability:(HPLC gradient) Passes test
Specification Test Results
Frequently Bought Together