ContactUsLinkComponent
Potassium hexacyanoferrate(II) trihydrate 99+% for analysis
Potassium hexacyanoferrate(II) trihydrate 99+% for analysis
Certificates
undefined
Potassium hexacyanoferrate(II) trihydrate 99+% for analysis
Specification Test Results
  • Appearance:
    Yellow Crystalline powder
  • Titration with KMnO4:
    ≥99.0 %
  • Chloride (Cl):
    ≤100 ppm
  • Sulfate (SO4):
    ≤100 ppm
  • Insoluble matter:
    ≤0.005 %