
Ammonium bromide ACS
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:12124-97-9
:
Formula:H₄BrN
MDL Number:MFCD00011419
:97.94 g/mol
Boiling Pt:235 °C (760 mmHg)
Melting Pt:452 °C (subl.)
:GHS07
Storage Temperature:Ambient
Specification Test Results
- Appearance:White Crystalline powder
- Sulfated ash:≤0.01 %
- Titration Argentimetric:≥99.0 %
- pH:4.5 to 6.0 (5 % at 25.0°C)
- Chloride (Cl):≤0.2 %
- Iodide (I):(Limit about 0.005 %) Passes test
- Bromate (BrO3):≤0.002 %
- Sulfate (SO4):≤0.005 %
- Barium (Ba):≤0.002 %
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Insoluble matter:≤0.005 %
Specification Test Results
Frequently Bought Together