Order Entry
United States
Orders LinkContactUsLinkComponent
 

Potassium nitrate 99%, pure

Potassium nitrate 99%, pure
 
Potassium nitrate 99%, pure
Potassium nitrate 99%, pure

 

 
   
 
 :  Nitrate of Potash
  •  :7757-79-1
  •  :KNO₃
  •  :MFCD00011409
  •  :101.1 g/mol
  •  :400 °C (760 mmHg)
  •  :334 °C
  •  :Ambient
  •  :1486
  •  :5.1,III
 

 

  • Appearance:
    White Crystalline powder and/or chunks
  • Loss on drying:
    ≤0.5 %
  • Titration after Ion exchange:
    ≥98.5 %
Product Family Options

 

  • Pack type
    Size
     
     
  • Glass bottle
    500 g
       
       
    Each (500g)
    Each (500g) - $38.30
    $38.30
    Each (500g)
       $38.30
     

     : AC44637-5000
  • Glass bottle
    2.5 kg
       
       
    Each (2,500g)
    Each (2,500g) - $149.00
    $149.00
    Each (2,500g)
       $149.00
     

     : AC44637-0025
  • Plastic drum
    10 kg
       
       
    Each (10kg)
    Each (10kg) - $320.00
    $320.00
    Each (10kg)
       $320.00

     : AC44637-0100

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.