Potassium acetate 99+% for biochemistry
Supplier: Thermo Scientific Chemicals
Certificates
About this item
CAS Number:127-08-2
:
Formula:CH₃COOK
MDL Number:MFCD00012458
:98.14 g/mol
Melting Pt:292 °C
Flash Pt:>250 °C
Storage Temperature:Ambient
Specification Test Results
- Appearance:Colorless to white Crystalline powder
- Infrared spectrum:Conforms
- Solubility:(5 M in water) Clear colorless solution
- pH:6.5 to 9.0
- Titration with HClO4:≥99.0 %
- Chloride (Cl):≤0.003 %
- Phosphate (PO4):≤0.001 %
- Sulfate (SO4):≤0.002 %
- Calcium (Ca):≤0.005 %
- Iron (Fe):≤0.0005 %
- Magnesium (Mg):≤0.002 %
- Sodium (Na):≤0.03 %
- Heavy metals (as Pb):≤5 ppm
Specification Test Results
Frequently Bought Together