Order Entry
United States
ContactUsLinkComponent
Sodium molybdate(VI) dihydrate 99+%
Sodium molybdate(VI) dihydrate 99+%
Certificates
undefined
Sodium molybdate(VI) dihydrate 99+%
Certificates

Specification Test Results

  • Appearance:
    White Crystalline powder
  • Assay:
    ≥99.0 %

Specification Test Results

Related Information

Frequently Bought Together