Imaliner® Low Absorption Polymer Lined Bi-layer Thermoplastic Elastomer (TPE) Tubing, Foxx Life Sciences
: Avantor
Imaliner® Bi-Layer Pharmaceutical Grade Thermoplastic Elastomer (TPE) tubing is specially designed with peristaltic pump compatibility in mind.
- Material: TPE
- Manufactured and packaged in Class 10000 cleanroom
- Suitable for in-situ sterilization
- Better peristaltic pump performance compared to Imaprene® tubing
- Low permeability
- Free of plasticizers (Phthalate/Bisphenol-A)
- Service temperature range: −40 to 135 °C
With excellent flex crack resistance, Imaliner® has a low coefficient of friction with a smooth bore to ensure less contamination and to enhance smooth fluid flow operation. The outer layer is made from abrasion-resistant, FDA compliant TPE to reduce spallation, while the inner layer is highly resistant to alcohol-based solvents and various preservatives (M-Cresol, Parabens, Benzyl Alcohol, BKC).
: FDA 2 CFR 177.2600. USP Class VI material. ISO 10993-11. TSE/BSE-free certification. Extraction study USFDA DMF #32559.
Frequently Bought Together