Order Entry
United States
ContactUsLinkComponent
Brady® Water Soluble Paper Labels with Ribbon for M210 M211 BMP21-PLUS Printers, Brady
Brady® Water Soluble Paper Labels with Ribbon for M210 M211 BMP21-PLUS Printers, Brady
Catalog # 76537-144
Supplier:  Brady Worldwide
undefined
Brady® Water Soluble Paper Labels with Ribbon for M210 M211 BMP21-PLUS Printers, Brady
Catalog # 76537-144
Supplier:  Brady Worldwide
Supplier Number:  M21-500-403

Specifications

  • For Use With:
    M210 Label Printer; M210-LAB Label Printer; M211 Label Printer
  • Color:
    Black on White
  • Length:
    25'
  • Material:
    Paper
  • Cat. No.:
    76537-144
  • Width:
    0.75"

Specifications

About this item

For the ultimate solution in label and adhesive removal, choose Brady’s B-403 Water Dissolvable Label. Ideal for laboratories, temporary identification needs, work-in-process product flows, and other general identification applications.

  • Acryclic adhesive
  • Matte finish
  • Dissolves in water

Warm water and 30 seconds is all it takes to completely remove the label and adhesive. From laboratories to restaurants, this dissolvable paper material is an excellent solution for temporary identification of any kind. Black print on white paper with easy to apply pressure-sensitive adhesive. For material specific performance and properties, please refer to the Technical Data Sheet (TDS).

Certifications: RoHS compliant to 2011/65/EU (Pass 2015/863).