Help ensure workers stay safe from hazardous energy during servicing and repair with strong, efficient, and cost-effective Safety Lockout Padlocks from Brady.
- Standard padlock body type
- Service temperature range: −15 to +250 °F
Designed exclusively for lockout tagout applications as part of an OSHA-compliant safety program. Key-retaining brass cylinder prevents key removal in the unlocked position for added safety and helps limit lost keys.
Nylon padlocks with steel shackles are lightweight, durable, and ideal for general lockout applications. Locks that are Keyed Different (KD) are ideal for simple lockout applications and a manageable number of energy isolation points. Record and retain engraved key codes to streamline replacement of lost keys. Locks that are Keyed Alike (KA) reduce the number of keys needed to carry and are ideal for individuals or trades responsible for multiple machines or isolation points.