Order Entry
United States
ContactUsLinkComponent
Human Recombinant GFRalpha1 (GFRA1)-Fc Chimera
Human Recombinant GFRalpha1 (GFRA1)-Fc Chimera
Supplier:  Biolegend
Certificates
undefined
Human Recombinant GFRalpha1 (GFRA1)-Fc Chimera
Supplier:  Biolegend

Specifications

  • Protein/Peptide Type:
    Recombinant
  • Source:
    CHO cells
  • Species:
    Human
  • Storage Conditions:
    –20…–70 °C
  • Carrier-Free:
    Y
  • Protein Synonyms:
    Glial cell line-derived neurotrophic factor receptor alpha|GDNFR|GDNFRA|GFRA1|GDNF family receptor alpha-1
  • Protein/Peptide Name:
    GFRalpha1 (GFRA1)-Fc Chimera
  • Concentration:
    0.2 mg/ml
  • Formulation:
    0.22 µm filtered protein solution is in PBS, pH 7.2
  • Tested Applications:
    BA

Specifications

Frequently Bought Together