Anti-CD14 Mouse Monoclonal Antibody [clone: 63D3] (Spark NIR™ 685)
Supplier: Biolegend
Certificates
About this item
Specifications
- Catalog No:
- 76422-628
- 76422-626
- Antigen Symbol:
- CD14
- CD14
- Antigen Synonyms:
- Monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein|LPS receptor
- Monocyte differentiation antigen CD14|myeloid cell-specific leucine-rich glycoprotein|LPS receptor
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- Spark NIR™ 685
- Spark NIR™ 685
- Clone:
- 63D3
- 63D3
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Purified human peripheral blood monocytes.
- Purified human peripheral blood monocytes.
- Purification:
- The antibody was purified by affinity chromatography and conjugated with Spark NIR™ 685 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with Spark NIR™ 685 under optimal conditions.
- Storage Buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA)
- Storage Temperature:
- 2...8 °C
- 2...8 °C
- Tested Applications:
- FC
- FC
Specifications
Frequently Bought Together