Anti-CD366 (Tim-3) Mouse Monoclonal Antibody [clone: F38-2E2] (PE (Phycoerythrin)/Cy5®)
- Catalog No:
- 76422-478
- 76422-480
- Antigen Symbol:
- CD366 (Tim-3)
- CD366 (Tim-3)
- Antigen Synonyms:
- hepatitis virus cellular receptor 2|T cell immunoglobulin and mucin domain containing protein 3|CD366
- hepatitis virus cellular receptor 2|T cell immunoglobulin and mucin domain containing protein 3|CD366
- Clonality:
- Monoclonal
- Monoclonal
- Conjugation:
- PE (Phycoerythrin)-Cy5®
- PE (Phycoerythrin)-Cy5®
- Clone:
- F38-2E2
- F38-2E2
- Reactivity:
- Human
- Human
- Host:
- Mouse
- Mouse
- Isotype:
- IgG1 kappa
- IgG1 kappa
- Flow Cytometry:
- Yes
- Yes
- Immunogen:
- Human Tim-3 fusion protein
- Human Tim-3 fusion protein
- Purification:
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine5 under optimal conditions.
- The antibody was purified by affinity chromatography and conjugated with PE/Cyanine5 under optimal conditions.
- Storage Buffer:
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 0.2% (w/v) BSA (origin USA).
- Storage Temperature:
- 2...8 °C
- 2...8 °C
- Tested Applications:
- FC
- FC
Frequently Bought Together