Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Chemical Structure
Manufacturer Data
0
$0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
United States
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Antibodies
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody [clone: 9E9A8] (TotalSeq™-C0597)
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody [clone: 9E9A8] (TotalSeq™-C0597)
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody [clone: 9E9A8] (TotalSeq™-C0597)
Catalog # 76422-552
Supplier:
Biolegend
Expand image
Anti-CD209 (DC-SIGN) Mouse Monoclonal Antibody [clone: 9E9A8] (TotalSeq™-C0597)
Catalog # 76422-552
Supplier:
Biolegend
Supplier Number:
330121
Specifications
Antigen Name:
DC-SIGN
Antigen Symbol:
CD209
Size:
10 µg
Cat. No.:
76422-552
Specifications
About this item
Product Details
Product Details