Anti-CD307e (FcRL5) Mouse Monoclonal Antibody [clone: 509f6] (TotalSeq™-C0829)
Catalog # 76422-230
Supplier: Biolegend
Specifications
- Antigen Symbol:CD307e (FcRL5)
- Clonality:Monoclonal
- Clone:509f6
- Conjugation:TotalSeq™-C0829
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Human
- Size:10 µg
- Antigen Synonyms:CD307|IRTA2|FcRH5|BXMAS1|IFGP5
- Storage Buffer:Phosphate-buffered solution, pH 7.2, containing 0.09% sodium azide and 1 mM EDTA
- Storage Temperature:2...8 °C
- Concentration:0.5 mg/ml
- Immunogen:P815 cells transfected with FcRL5
- Tested Applications:PG
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-C oligomer under optimal conditions.
- Cat. No.:76422-230