Anti-CD62P (P-selectin) Mouse Monoclonal Antibody [clone:RMP-1] (TotalSeq™-B0229)
Catalog # 76508-932
Supplier: Biolegend
Specifications
- Antigen Symbol:CD62P (P-selectin)
- Clonality:Monoclonal
- Clone:RMP-1
- Conjugation:TotalSeq™-B0229
- Host:Mouse
- Isotype:IgG2a kappa
- Reactivity:Rat,Mouse
- Size:10 µg
- Antigen Synonyms:GMP-140|Platelet activation-dependent granule membrane protein (PADGEM)
- Storage Temperature:2...8 °C
- Concentration:0.5 mg/ml
- Immunogen:Thrombin activated rat platelets
- Purification:The antibody was purified by chromatography and conjugated with TotalSeq™-B oligomer under optimal conditions.
- Cat. No.:76508-932