Order Entry
United States
ContactUsLinkComponent
Sodium dodecyl sulfate (SDS) 200 g/l in aqueous solution
Sodium dodecyl sulfate (SDS) 200 g/l in aqueous solution
Catalog # 101643-244
Supplier:  Bio-Rad
CAS Number:  
Sodium dodecyl sulfate (SDS) 200 g/l in aqueous solution
Catalog # 101643-244
Supplier:  Bio-Rad
Supplier Number:  1610418
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Pack type:
    Glass bottle
  • Size:
    1 L
  • Cat. No.:
    101643-244
  • Supplier no.:
    1610418

Specifications

About this item

 :
 :288.38 g/mol
Density:1…1.1 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-788-1
CAS Number:151-21-3
Formula:CH₃(CH₂)₁₁OSO₃Na
MDL Number:MFCD00036175