ContactUsLinkComponent
Cell Culture Media, Powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Cell Culture Media, Powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Catalog # 76461-570
Supplier:  MP Biomedicals
CAS Number:  
Cell Culture Media, Powdered, Basal Medium Eagle (BME) Modified, MP Biomedicals
Catalog # 76461-570
Supplier:  MP Biomedicals
Supplier Number:  091003122
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Media Format:
    Powder
  • Description:
    BME (Modified), with Hanks' salts and L-glutamine, without sodium bicarbonate, 1×
    10 l
  • Cat. No.:
    76461-570
Specifications
About this item
Basal Medium Eagle is suited for the isolation and cultivation of non-fastidious and fastidious microorganisms from a variety of clinical and non-clinical specimens. It can also be used as a selective isolation medium by adding antimicrobial agents.