ContactUsLinkComponent
arium® comfort Pre-treatment Cartridge, Sartorius
arium® comfort Pre-treatment Cartridge, Sartorius
Catalog # 76533-172
Supplier:  Sartorius
arium® comfort Pre-treatment Cartridge, Sartorius
Catalog # 76533-172
Supplier:  Sartorius
Supplier Number:  H2O-CPFCO-1
Specifications
  • Description:
    arium® comfort pre-treatment cartridge
  • W×D×H:
    280×210×140 mm
  • Cat. No.:
    76533-172
Specifications
About this item
Pre-filter consisting of high efficient activated carbon combined with a process optimizing catalyst. Removes reliable oxidizing agents, heavy metals, and particular impurities.
  • Fast and effective adsorption of impurities through high-grade activated carbon
  • Highly efficient catalyst for removing free chlorine
The combination of spherical, catalytically active activated carbon with an added catalyst constitutes reliable protection for a downstream reverse osmosis (RO) membrane. The cartridge removes oxidation agents, such as free chlorine and ozone, heavy metal ions, and particulate contaminants from the feed water of the system.
This cartridge is designed specifically for use with arium® comfort I and arium® comfort II systems.
Materials of construction: High-quality polypropylene