ContactUsLinkComponent
Pipette/Instrument Organizer Tray, United Scientific
Pipette/Instrument Organizer Tray, United Scientific
Catalog # 470332-806
undefined
Pipette/Instrument Organizer Tray, United Scientific
Catalog # 470332-806
Supplier Number:  PPT004
Specifications
  • Description:
    Pipette/Instrument organizer tray
  • Cat. No.:
    470332-806
Specifications
About this item
Durable polypropylene tray for organizing pipettes or other instruments.
  • Tray measures 16×12×1"
  • Ideal for drawers, refrigerators or bench tops
  • Tray walls are ¹/₈" thick
  • Autoclavable
The pipette tray is designed to fit large drawers, refrigerators, and benchtops. Tray features 4 deep columns (approx. 0.75" deep) and is recommended for 1 to 10 ml pipettes. Molded-in polypropylene with ¹/₈" thick walls.