Newton's Ring Apparatus, United Scientific
Catalog # 470332-746
Supplier: United Scientific Supplies
Specifications
- Description:Newton's ring apparatus
- Cat. No.:470332-746
Specifications
About this item
The Newton’s Rings Apparatus consists of a plano-convex lens on top of a flat glass plate, held together by a plastic frame.
- Easy to adjust thumbscrews
- Demonstrate optical properties of lenses
The pressure between the lens and the plate can be adjusted by means of three thumbscrews.