Loop The Loop Track, United Scientific
Catalog # 470332-770
Supplier: United Scientific Supplies
Specifications
- Description:Loop the loop track
- Cat. No.:470332-770
Specifications
About this item
Ideal for classroom demonstrations of centripetal acceleration and transformation of potential energy into kinetic energy.
- Includes activity guide
The Loop the Loop Track demonstrates centripetal acceleration and the transformation of potential energy into kinetic energy. Included is a sturdy base with rubber feet, metal ramp with loop, 16 mm steel ball, and 19 mm steel ball. The metal ramp with loop features a net at the end of the loop to receive the steel ball.