Orders LinkContactUsLinkComponent
Solar Kit, Basic, United Scientific
Solar kit, basic
  470332-754
 :  SLRKIT-B
Solar Kit, Basic, United Scientific
  470332-754
Solar Kit, Basic, United Scientific
Solar Kit, Basic, United Scientific
Solar kit, basic
$10.70
Each
Convenient kit provides an introduction to the conversion of sunlight into electrical energy.
  • Includes small solar panel and three energy consumers: buzzer, LED, and motor with fan
  • Mirror can be used to observe angle of incidence and its impact on energy generation
  • Activity guide also included
The Solar Kit offers a convenient introduction to the conversion of solar light energy into electrical energy. The conversion is carried out by a photoelectric panel, and three 'consumers' are included to demonstrate the use of the electrical energy; an LED shows the conversion back to light, a buzzer shows the conversion to sound energy, and a motor demonstrates the conversion to mechanical energy. A mirror is also included to allow various amounts of sunlight to be reflected onto the panel to investigate the effect of the amount of solar energy on the electrical energy produced.
Kit includes solar panel, buzzer, red LED, motor with fan, mirror, and activity guide.
  • Description:
    Solar kit, basic