Test Tube Racks, Caplugs
Supplier: Caplugs
Certificates
About this item
Caplugs Evergreen Test tube racks are easy to assemble and available for use with common test tube sizes.
- 240-3200, 240-3201, and 240-3204 series
- Lightweight, stackable, and autoclavable
- Racks fit 13, 16, or 30 mm test tubes
- All racks measure 246×104×64 mm
Racks are submersible in water and resistant to common organic chemicals found in laboratories such as ethanol, NBF (neutral buffered formain), and xylene.
Frequently Bought Together