Beakers, Berzelius, Tall Form, Borosilicate Glass, United Scientific Supplies
Catalog # 76531-410
Supplier: United Scientific Supplies
Supplier #: BG1060-400
Log in for availability
About this item
Made from low-expansion borosilicate glass, these beakers feature heavy and uniform wall thickness, spouts designed for easy pouring, and beaded tops.
- Feature beaded rims
- Include white printed graduations (from 20 - 80 ml, at 10 ml intervals) and durable marking area
- Tolerance is approximately +/- 5%
Available in multiple sizes.
Frequently Bought Together