Order Entry
United States
ContactUsLinkComponent
Cell Meter™ Fluorimetric Cellular Lipid Peroxidation Assay Kit, AAT Bioquest
Cell Meter™ Fluorimetric Cellular Lipid Peroxidation Assay Kit, AAT Bioquest
Catalog # 76484-744
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ Fluorimetric Cellular Lipid Peroxidation Assay Kit, AAT Bioquest
Catalog # 76484-744
Supplier:  AAT BIOQUEST INC
Supplier Number:  22906
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    Cell Meter™ Fluorimetric cellular lipid peroxidation assay kit
  • Size:
    200 Tests
  • Cat. No.:
    76484-744
  • No. of tests:
    200

Specifications

About this item

Lipid peroxidation is the oxidative degradation of cellular lipid by reactive oxygen species (ROS).

  • Contains all necessary components
  • Multiple research applications
  • Ratiometric detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.