Cell Meter™ Intracellular GSH Assay Kit *Optimized for Flow Cytometry*, AAT Bioquest
Catalog # 76483-764
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Intracellular GSH assay kit *optimized for flow cytometry*
- Size:100 Tests
- Cat. No.:76483-764
- No. of tests:100
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.