Order Entry
United States
ContactUsLinkComponent
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*, AAT Bioquest
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*, AAT Bioquest
Catalog # 76483-758
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ NIR Mitochondrion Membrane Potential Assay Kit *Optimized for Microplate Reader*, AAT Bioquest
Catalog # 76483-758
Supplier:  AAT BIOQUEST INC
Supplier Number:  22803
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    Cell Meter™ NIR mitochondrion membrane potential assay kit *optimized for microplate reader*
  • Size:
    500 Tests
  • Cat. No.:
    76483-758
  • No. of tests:
    500

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Optimized for microplate readers but suitable for all major fluorescence platforms
Caution: For research use only.