Order Entry
United States
ContactUsLinkComponent
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimized for Microplate Readers*, AAT Bioquest
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimized for Microplate Readers*, AAT Bioquest
Catalog # 76483-736
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Red Fluorescence Optimized for Microplate Readers*, AAT Bioquest
Catalog # 76483-736
Supplier:  AAT BIOQUEST INC
Supplier Number:  22792
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Specifications

  • Description:
    Cell Meter™ Phosphatidylserine apoptosis assay kit *red fluorescence optimized for microplate readers*
  • Size:
    100 Tests
  • Cat. No.:
    76483-736
  • No. of tests:
    100

Specifications

About this item

Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.

  • Contains all necessary components
  • Multiple research applications
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.