Order Entry
United States
ContactUsLinkComponent
TRITC-Dextran Conjugate (Average MW = ~70K), AAT Bioquest
TRITC-Dextran Conjugate (Average MW = ~70K), AAT Bioquest
Catalog # 76485-180
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
TRITC-Dextran Conjugate (Average MW = ~70K), AAT Bioquest
Catalog # 76485-180
Supplier:  AAT BIOQUEST INC
Supplier Number:  21708
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    TRITC–dextran conjugate (average MW = ~70K)
  • Size:
    25 mg
  • Cat. No.:
    76485-180

Specifications

About this item

TRITC-labeled dextran is used in cardiovascular, microcirculation, perfusion, cell monolayer and cell membrane permeability research as fluorescent flux tracer compound that supports the measurement of processes such as blood flow, membrane damage, vascular drainage and renal elimination.

  • High quality
  • Multiple applications
Caution: For research use only.