Screen Quest™ Fluorimetric Glucose Uptake Assay Kit, AAT Bioquest
Supplier: AAT Bioquest
Certificates
About this item
Glucose transport systems are responsible for transporting glucose across cell membranes.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for microplate reader but suitable for all major fluorescence platforms
Caution: For research use only.
Frequently Bought Together