Order Entry
United States
ContactUsLinkComponent
 
Cell Meter™ Fluorimetric Cellular Voltage Assay Kit, AAT Bioquest
Catalog # 76485-486
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ Fluorimetric Cellular Voltage Assay Kit, AAT Bioquest
Catalog # 76485-486
Supplier:  AAT BIOQUEST INC
Supplier Number:  35000
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Cell Meter™ Fluorimetric cellular voltage assay kit
  • Size:
    100 Tests
  • Cat. No.:
    76485-486
  • No. of tests:
    100

Specifications

About this item

Almost all plasma membranes have an electrical potential across them, with the inside usually negative with respect to the outside.

  • Contains all necessary components
  • Multiple research applications
  • Ratiometric detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.