Cell Meter™ Fluorimetric Fixed Cell Cycle Assay Kit *Red Fluorescence Optimized for Flow Cytometry*, AAT Bioquest
Catalog # 76483-814
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Fluorimetric fixed cell cycle assay kit *Red fluorescence optimized for flow cytometry*
- Size:100 Tests
- Cat. No.:76483-814
- No. of tests:100
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability and proliferation.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.