Cell Meter™ Generic Fluorimetric Caspase Activity Assay Kit *Green Fluorescence Optimized for Flow Cytometry*, AAT Bioquest
Catalog # 76483-778
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Generic fluorimetric caspase activity assay kit *Green fluorescence optimized for flow cytometry*
- Size:100 Tests
- Cat. No.:76483-778
- No. of tests:100
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for flow cytometry but suitable for all major fluorescence platforms
Caution: For research use only.