Cell Meter™ Mitochondrion Membrane Potential Assay Kit *Red Fluorescence Optimized for Microplate Reader*, AAT Bioquest
Catalog # 76484-670
Supplier: AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.
Specifications
- Description:Cell Meter™ Mitochondrion membrane potential assay kit *Red fluorescence optimized for microplate reader*
- Size:500 Tests
- Cat. No.:76484-670
- No. of tests:500
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Optimized for microplate readers but suitable for all major fluorescence platforms
Caution: For research use only.