Cell Meter™ Cell Viability Assay Kit *Green Fluorescence*, AAT Bioquest
Catalog # 76483-724
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ Cell viability assay kit *Green fluorescence*
- Size:500 Tests
- Cat. No.:76483-724
- No. of tests:500
Specifications
About this item
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
- Results in under 2 hours
Caution: For research use only.