ContactUsLinkComponent
Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*, AAT Bioquest
Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*, AAT Bioquest
Catalog # 76484-486
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Navigator® Fluorimetric Lipid Droplet Assay Kit *Red Fluorescence*, AAT Bioquest
Catalog # 76484-486
Supplier:  AAT BIOQUEST INC
Supplier Number:  22735
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cell Navigator™ Fluorimetric lipid droplet assay kit *Red fluorescence*
  • Size:
    200 Tests
  • Cat. No.:
    76484-486
  • No. of tests:
    200
Specifications
About this item
Lipid droplets, also referred to as lipid bodies, oil bodies or adiposomes, are lipid-rich cellular organelles that regulate the storage and hydrolysis of neutral lipids.
  • Live cell labeling
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.