ContactUsLinkComponent
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence*, AAT Bioquest
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence*, AAT Bioquest
Catalog # 76483-686
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Navigator® Mitochondrion Staining Kit *Orange Fluorescence*, AAT Bioquest
Catalog # 76483-686
Supplier:  AAT BIOQUEST INC
Supplier Number:  22667
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 
Specifications
  • Description:
    Cell Navigator™ Mitochondrion staining kit *Orange fluorescence*
  • Size:
    500 Tests
  • Cat. No.:
    76483-686
Specifications
About this item
Our Cell Navigator® fluorescence imaging kits are a set of fluorescence imaging tools for labeling sub-cellular organelles such as membranes, lysosomes, mitochondria and nuclei etc.
  • Cell-permeant staining
  • Improved signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.