Order Entry
United States
ContactUsLinkComponent
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*, AAT Bioquest
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*, AAT Bioquest
Catalog # 76483-628
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Explorer™ Live Cell Labeling Kit *Orange Fluorescence with 405 nm Excitation*, AAT Bioquest
Catalog # 76483-628
Supplier:  AAT BIOQUEST INC
Supplier Number:  22616
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Cell Explorer™ Live Cell labeling kit *Orange fluorescence with 405 nm excitation*
  • Size:
    200 Tests
  • Cat. No.:
    76483-628
  • No. of tests:
    200

Specifications

About this item

Our Cell Explorer™ fluorescence imaging kits are a set of tools for labeling cells for fluorescence microscopic investigations of cellular functions.

  • Contains all necessary components
  • Multiple research applications
  • Large Stokes shift
  • Bright and stable signal
  • Suitable for all major fluorescence platforms
Caution: For research use only.