ContactUsLinkComponent
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*, AAT Bioquest
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*, AAT Bioquest
Catalog # 76483-444
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
PhosphoWorks™ Colorimetric MESG Phosphate Assay Kit *UV Absorption*, AAT Bioquest
Catalog # 76483-444
Supplier:  AAT BIOQUEST INC
Supplier Number:  21659
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    PhosphoWorks™ Colorimetric MESG phosphate assay kit *UV absorption*
  • Size:
    200 Tests
  • Cat. No.:
    76483-444
  • No. of tests:
    200
Specifications
About this item
In the presence of inorganic phosphate MESG is converted to 2-amino-6-mercapto-7-methlpurine by purine nucleoside phosphorylase (EC 2.4.2.1) with absorption wavelength shift to red.
  • Provides all necessary components
  • Multiple applications
  • Photometric detection
Caution: For research use only.