Order Entry
United States
ContactUsLinkComponent
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*, AAT Bioquest
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*, AAT Bioquest
Catalog # 76484-568
Supplier:  AAT Bioquest
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit *Optimized for microplate reader*, AAT Bioquest
Catalog # 76484-568
Supplier:  AAT Bioquest
Supplier Number:  36312
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  

Some Products May Appear Restricted

To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.

If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/us/en/login/register)

 

If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation;  additional documentation will be requested for these items prior to shipment. 

Specifications

  • Description:
    Cell Meter™ no wash and probenecid-free endpoint calcium assay kit *optimized for microplate reader*
  • Size:
    100 Tests
  • Cat. No.:
    76484-568
  • No. of tests:
    100

Specifications

About this item

Cell Meter™ No Wash and Probenecid-Free Endpoint Calcium Assay Kit enables homogeneous fluorescence-based assays for detecting intracellular calcium mobilization without the need to use kinetics reading mode.

  • Ready to use
  • Contains all necessary components
  • Robust and adaptable
  • Optimized for microplate readers but suitable for all major fluorescence platforms
  • Multiple research applications
Caution: For research use only.