Cell Meter™ Autophagy Fluorescence Imaging Kit, AAT Bioquest
Catalog # 76483-840
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ autophagy fluorescence Imaging kit
- Size:200 Tests
- Cat. No.:76483-840
- No. of tests:200
Specifications
About this item
Autophagy is one of the major pathways for degradation of intracellular macromolecules in animal cells.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.