Cell Meter™ Caspase 3/7 Activity Apoptosis Assay Kit *Red Fluorescence*, AAT Bioquest
76483-746
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
:
- Description:Cell Meter™ caspase ³/₇ activity apoptosis assay kit *red fluorescence*
- Size:100 Tests
- Cat. No.:76483-746
- No. of tests:100
Our Cell Meter™ assay kits are a set of tools for monitoring cell viability.
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
: For research use only.