Cell Meter™ Phosphatidylserine Apoptosis Assay Kit *Orange Fluorescence Optimized for Microplate Readers*, AAT Bioquest
Catalog # 76483-740
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:Cell Meter™ phosphatidylserine apoptosis assay kit *orange fluorescence optimized for microplate readers*
- Size:100 Tests
- Cat. No.:76483-740
- No. of tests:100
Specifications
About this item
This particular kit is designed to monitor cell apoptosis through measuring the translocation of phosphatidylserine (PS).
- Contains all necessary components
- Multiple research applications
- Bright and stable signal
- Suitable for all major fluorescence platforms
Caution: For research use only.