Specifications
- Description:Cell Navigator™ cell plasma membrane staining kit *green fluorescence*
- Size:500 Tests
- Cat. No.:76485-046
- No. of tests:500
Specifications
About this item
The Cell Navigator® Plasma Membrane Stain Kit provides a fast and uniform labeling of the plasma membrane without the cell-type differences exhibited by lectins.
- Signal persists through fixation
- Stable labeling regardless of cell type
- Multiple applications