PhosphoWorks™ Fluorimetric Pyrophosphate Assay Kit, Blue Fluorescence, AAT Bioquest
Catalog # 76483-434
Supplier: AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:
Specifications
- Description:PhosphoWorks™ fluorimetric pyrophosphate assay kit *blue fluorescence*
- Size:200 Tests
- Cat. No.:76483-434
- No. of tests:200
Specifications
About this item
Pyrophosphate (PPi) is produced by a number of biochemical reactions, such as ATP hydrolysis, DNA and RNA polymerizations, cyclic AMP formation by the enzyme adenylate cyclase and the enzymatic activation of fatty acids to form their coenzyme A esters.
- Pyrophosphate-dependent fluorescence
- Highly selective for PPi
- Contains all necessary components
Caution: For research use only.