Specifications
- Description:Cell Meter™ Live cell Caspase 3/7 Imaging kit *Green fluorescence*
- Size:100 tests
- Cat. No.:76485-692
- No. of tests:100
Specifications
About this item
Cell Meter™ Live Cell No Wash Caspase 3/7 Imaging Kit uses our recently developed cell-permeable fluorogenic caspase substrate, ApoSight™ Green Caspase 3/7, the first fluorogenic probe, for the direct detection of caspase activities in live cells.
- Based on FMK inhibitor
- Direct caspase detection
- Suitable for all major fluorescence platforms