ContactUsLinkComponent
Cell Meter™ Live Cell Caspase 6 Binding Assay Kit *Green Fluorescence*, AAT Bioquest
Cell Meter™ Live Cell Caspase 6 Binding Assay Kit *Green Fluorescence*, AAT Bioquest
Catalog # 76483-032
Supplier:  AAT BIOQUEST INC
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
undefined
Cell Meter™ Live Cell Caspase 6 Binding Assay Kit *Green Fluorescence*, AAT Bioquest
Catalog # 76483-032
Supplier:  AAT BIOQUEST INC
Supplier Number:  20113
Minority or Woman-Owned Business Enterprise
Small Business Enterprise
CAS Number:  
Specifications
  • Description:
    Cell Meter™ Live cell Caspase 6 binding assay kit *Green fluorescence*
  • Size:
    25 tests
  • Cat. No.:
    76483-032
  • No. of tests:
    25
Specifications
About this item
Our Cell Meter™ live cell caspases activity assay kits are based on fluorescent FMK inhibitors of caspases.
  • Based on FMK inhibitor
  • Direct caspase detection
  • Suitable for all major fluorescence platforms
Caution: For research use only.